2,3-Difluor-6-nitrophenol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,2-Difluoro-3-hydroxy-4-nitrobenzene

PC0285-25G PC0285-10G PC0285-1G
APOSPC0285-25GEA 31.9 EUR
APOSPC0285-25G APOSPC0285-10G APOSPC0285-1G
2,3-Difluor-6-nitrophenol 97%
2,3-Difluor-6-nitrophenol
Formel: C₆H₃F₂NO₃
Molecular Weight: 175.09 g/mol
Schmelzpunkt: 60…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042255
CAS-Nummer: 82419-26-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR