Boc-3,4,5-trifluoro-D-phenylalanine

Lieferant: Apollo Scientific

Synonyme: (R)-2-((tert-Butoxycarbonyl)amino)-3-(3,4,5-trifluorophenyl)propanoic acid, Boc-D-Phe(3,4,5-DiF)-OH

PC0309-1G PC0309-250MG PC0309-5G
APOSPC0309-1GEA 256 EUR
APOSPC0309-1G APOSPC0309-250MG APOSPC0309-5G
Boc-3,4,5-trifluoro-D-phenylalanine
Boc-3,4,5-trifluoro-D-phenylalanine
Formel: C₁₄H₁₆F₃NO₄
Lagertemperatur: Kühlschrank
MDL: MFCD00797559
CAS-Nummer: 205445-55-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR