2-Chlor-4-nitrobenzotrifluorid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chlor-4-nitrobenzotrifluorid, 1-Chlor-3-nitro-6-(trifluormethyl)benzol

PC0322-1G PC0322-5G
APOSPC0322-1GEA 21.9 EUR
APOSPC0322-1G APOSPC0322-5G
2-Chlor-4-nitrobenzotrifluorid
2-Chlor-4-nitrobenzotrifluorid
Formel: C₇H₃ClF₃NO₂
Molecular Weight: 225.55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00171532
CAS-Nummer: 151504-80-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR