3-Chlor-5-fluorphenylacetonitril

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Chloro-5-fluorophenylacetonitrile, 2-(3-Chloro-5-fluorophenyl)acetonitrile, 3-Chloro-5-fluorobenzyl cyanide

PC0331-1G PC0331-5G PC0331-25G
APOSPC0331-1GEA 24.8 EUR
APOSPC0331-1G APOSPC0331-5G APOSPC0331-25G
3-Chlor-5-fluorphenylacetonitril
3-Chlor-5-fluorphenylacetonitril
Formel: C₈H₅ClFN
Molecular Weight: 169,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038259
CAS-Nummer: 493038-93-0
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR