4-Fluoro-3-methylbenzamide

Lieferant: Apollo Scientific

PC0344-10G PC0344-5G PC0344-1G
APOSPC0344-10GEA 282 EUR
APOSPC0344-10G APOSPC0344-5G APOSPC0344-1G
4-Fluoro-3-methylbenzamide
4-Fluoro-3-methylbenzamide
Formel: C₈H₈FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD01631489
CAS-Nummer: 261945-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR