Fmoc-D-Pentafluorophenylalanine
Lieferant: Apollo Scientific
Synonyme:
Fmoc-D-Phe(F5)-OH, Fmoc-D-Phe(F)₅-OH
PC0348-1G
PC0348-5G
APOSPC0348-1GEA
150
EUR
APOSPC0348-1G
APOSPC0348-5G
Fmoc-D-Pentafluorophenylalanine
Fmoc-D-Pentafluorophenylalanine
Formel:
C₂₄H₁₆F₅NO₄ Molecular Weight: 477,39 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00672558 CAS-Nummer: 198545-85-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...