4-Fluor-3-methylbenzylalkohol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Fluoro-3-methylphenyl)methanol
PC0352-10G
PC0352-5G
PC0352-1G
APOSPC0352-10GEA
184
EUR
APOSPC0352-10G
APOSPC0352-5G
APOSPC0352-1G
4-Fluor-3-methylbenzylalkohol 98%
4-Fluor-3-methylbenzylalkohol
Formel:
C₈H₉FO Molecular Weight: 140,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631427 CAS-Nummer: 261951-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...