3-Fluor-4-methylbenzylbromid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-(Bromomethyl)-2-fluoro-1-methylbenzene
PC0360-10G
PC0360-50G
PC0360-5G
APOSPC0360-10GEA
116
EUR
APOSPC0360-10G
APOSPC0360-50G
APOSPC0360-5G
3-Fluor-4-methylbenzylbromid
3-Fluor-4-methylbenzylbromid
Formel:
C₈H₈BrF Molecular Weight: 203,05 g/mol Siedepunkt: 77 °C/3mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631534 CAS-Nummer: 145075-44-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...