(3-Fluor-4-methylphenyl)essigsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(3-Fluoro-4-methylphenyl)acetic acid

PC0370-250MG PC0370-1G PC0370-5G PC0370-10G
APOSPC0370-250MGEA 0 EUR
APOSPC0370-250MG APOSPC0370-1G APOSPC0370-5G APOSPC0370-10G
(3-Fluor-4-methylphenyl)essigsäure
(3-Fluor-4-methylphenyl)essigsäure
Formel: C₉H₉FO₂
Molecular Weight: 168,17 g/mol
Schmelzpunkt: 104…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631536
CAS-Nummer: 261951-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR