(5-Fluor-2-methylphenyl)essigsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC0371-1G
PC0371-5G
PC0371-25G
APOSPC0371-1GEA
50.9
EUR
APOSPC0371-1G
APOSPC0371-5G
APOSPC0371-25G
(5-Fluor-2-methylphenyl)essigsäure 97%
(5-Fluor-2-methylphenyl)essigsäure
Formel:
C₉H₉FO₂ Molecular Weight: 168,17 g/mol Schmelzpunkt: 120…123 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631530 CAS-Nummer: 261951-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...