4-Fluoro-3-methyl-DL-phenylglycine

Lieferant: Apollo Scientific

PC0376-1G PC0376-5G PC0376-10G
APOSPC0376-1GEA 116 EUR
APOSPC0376-1G APOSPC0376-5G APOSPC0376-10G
4-Fluoro-3-methyl-DL-phenylglycine
4-Fluoro-3-methyl-DL-phenylglycine
Formel: C₉H₁₀FNO₂
Molecular Weight: 183.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631428
CAS-Nummer: 261951-77-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR