4-Fluoro-DL-phenylglycine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluoro-DL-α-phenylglycine, 4-Fluorophenylglycine, H-DL-Phg(4-F)-OH

PC0378-5G PC0378-100G PC0378-25G
APOSPC0378-5GEA 21.9 EUR
APOSPC0378-5G APOSPC0378-100G APOSPC0378-25G
4-Fluoro-DL-phenylglycine 98%
4-Fluoro-DL-phenylglycine
Formel: C₈H₈FNO₂
Molecular Weight: 169.155 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00066444
CAS-Nummer: 7292-73-1
EINECS: 230-724-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR