4-Methoxy-3-(trifluormethyl)benzoylchlorid

Lieferant: Apollo Scientific
Gefahr

PC0407-1G PC0407-5G
APOSPC0407-1GEA 33.5 EUR
APOSPC0407-1G APOSPC0407-5G
4-Methoxy-3-(trifluormethyl)benzoylchlorid
4-Methoxy-3-(trifluormethyl)benzoylchlorid
Formel: C₉H₆ClF₃O₂
Molecular Weight: 238,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631576
CAS-Nummer: 98187-18-9
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR