4-Methoxy-3-(trifluormethyl)benzylbromid

Lieferant: Apollo Scientific
Gefahr

PC0411-10G PC0411-5G
APOSPC0411-10GEA 337 EUR
APOSPC0411-10G APOSPC0411-5G
4-Methoxy-3-(trifluormethyl)benzylbromid
4-Methoxy-3-(trifluormethyl)benzylbromid
Formel: C₉H₈BrF₃O
Molecular Weight: 269.06 g/mol
Schmelzpunkt: 75…79 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631578
CAS-Nummer: 261951-89-7
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR