2-Methyl-4-(trifluormethoxy)benzamid 98%

Lieferant: Apollo Scientific

PC0415-1G PC0415-5G
APOSPC0415-1GEA 372 EUR
APOSPC0415-1G APOSPC0415-5G
2-Methyl-4-(trifluormethoxy)benzamid 98%
2-Methyl-4-(trifluormethoxy)benzamid
Formel: C₉H₈F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01631494
CAS-Nummer: 261951-90-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR