[2-Methyl-4-(trifluoromethoxy)phenyl]methanol 97%

Lieferant: Apollo Scientific
Achtung

PC0420-1G PC0420-5G
APOSPC0420-1GEA 132 EUR
APOSPC0420-1G APOSPC0420-5G
[2-Methyl-4-(trifluoromethoxy)phenyl]methanol 97%
[2-Methyl-4-(trifluoromethoxy)phenyl]methanol
Formel: C₉H₉F₃O₂
Molecular Weight: 206,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631546
CAS-Nummer: 261951-94-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR