1-Brom-2-methyl-4-(trifluormethoxy)benzol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-5-(trifluoromethoxy)toluene
PC0433-10G
PC0433-1G
PC0433-5G
APOSPC0433-10GEA
94.4
EUR
APOSPC0433-10G
APOSPC0433-1G
APOSPC0433-5G
1-Brom-2-methyl-4-(trifluormethoxy)benzol 98%
1-Bromo-2-methyl-4-(trifluoromethoxy)benzene
Formel:
C₈H₆BrF₃O Molecular Weight: 255,0319 g/mol Dichte: 1,559 g/cm³ (25 °C) Flammpunkt: 78,3 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631545 CAS-Nummer: 261951-96-6 UN: 3082 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...