3-Amino-5-methylbenzotrifluorid 97%

Lieferant: Apollo Scientific
Gefahr

PC0435-1G PC0435-5G
APOSPC0435-1GEA 135 EUR
APOSPC0435-1G APOSPC0435-5G
3-Amino-5-methylbenzotrifluorid 97%
3-Amino-5-methylbenzotrifluorid
Formel: C₈H₈F₃N
Molecular Weight: 175,1510 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631580
CAS-Nummer: 96100-12-8
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR