N-(1-Benzyl-3-pyrrolidinyl)-2,2,2-trifluoracetamid
Lieferant: Apollo Scientific
|
Achtung
|
PC0436-1G
PC0436-5G
APOSPC0436-1GEA
27.7
EUR
APOSPC0436-1G
APOSPC0436-5G
N-(1-Benzyl-3-pyrrolidinyl)-2,2,2-trifluoracetamid
1-Benzyl-3-(trifluoroacetamido)pyrrolidine
Formel:
C₁₃H₁₅F₃N₂O Molecular Weight: 272,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059038 CAS-Nummer: 115445-23-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...