2-Methyl-5-(trifluormethyl)benzamid 97%

Lieferant: Apollo Scientific

PC0441-10G PC0441-5G PC0441-1G
APOSPC0441-10GEA 260 EUR
APOSPC0441-10G APOSPC0441-5G APOSPC0441-1G
2-Methyl-5-(trifluormethyl)benzamid 97%
2-Methyl-5-(trifluormethyl)benzamid
Formel: C₉H₈F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD01631496
CAS-Nummer: 261951-97-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR