4-Methyl-3-(trifluormethyl)benzamid

Lieferant: Apollo Scientific
Achtung

PC0444-10G PC0444-1G PC0444-5G
APOSPC0444-10GEA 786 EUR
APOSPC0444-10G APOSPC0444-1G APOSPC0444-5G
4-Methyl-3-(trifluormethyl)benzamid
4-Methyl-3-(trifluormethyl)benzamid
Formel: C₉H₈F₃NO
Lagertemperatur: Raumtemperatur
CAS-Nummer: 261952-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR