3-Methyl-5-(trifluormethyl)benzoesäure 99%

Lieferant: Apollo Scientific
Achtung

PC0448-1G PC0448-5G
APOSPC0448-1GEA 109 EUR
APOSPC0448-1G APOSPC0448-5G
3-Methyl-5-(trifluormethyl)benzoesäure 99%
3-Methyl-5-(trifluoromethyl)benzoic acid
Formel: C₉H₇F₃O₂
Molecular Weight: 204.15 g/mol
Schmelzpunkt: 127…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631590
CAS-Nummer: 117186-02-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR