Cyclopropyldiphenylsulfonium-tetrafluoroborat

Lieferant: Apollo Scientific

PC0457-1G PC0457-5G
APOSPC0457-1GEA 84.1 EUR
APOSPC0457-1G APOSPC0457-5G
Cyclopropyldiphenylsulfonium-tetrafluoroborat
Cyclopropyldiphenylsulphonium tetrafluoroborate
Formel: C₁₅H₁₅BF₄S
Molecular Weight: 314.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011873
CAS-Nummer: 33462-81-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR