2-Methyl-3-(trifluormethyl)benzylbromid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(Bromomethyl)-2-methyl-3-(trifluoromethyl)benzene
PC0479-1G
PC0479-5G
PC0479-10G
PC0479-25G
PC0479-100G
APOSPC0479-1GEA
0
EUR
APOSPC0479-1G
APOSPC0479-5G
APOSPC0479-10G
APOSPC0479-25G
APOSPC0479-100G
2-Methyl-3-(trifluormethyl)benzylbromid 98%
2-Methyl-3-(trifluormethyl)benzylbromid
Formel:
C₉H₈BrF₃ Molecular Weight: 253,06 g/mol Siedepunkt: 72 °C/1mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631608 CAS-Nummer: 261952-16-3 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...