3-Brom-2-methylbenzotrifluorid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-2-methyl-3-(trifluoromethyl)benzene
PC0486-1G
PC0486-25G
PC0486-5G
APOSPC0486-1GEA
21.9
EUR
APOSPC0486-1G
APOSPC0486-25G
APOSPC0486-5G
3-Brom-2-methylbenzotrifluorid 97%
3-Brom-2-methylbenzotrifluorid
Formel:
C₈H₆BrF₃ Molecular Weight: 239,04 g/mol Siedepunkt: 35 °C/1mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038231 CAS-Nummer: 69902-83-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...