3-(Trifluoromethoxy)-DL-phenylglycine
Lieferant: Apollo Scientific
|
Achtung
|
PC0517-1G
PC0517-5G
PC0517-250MG
APOSPC0517-1GEA
192
EUR
APOSPC0517-1G
APOSPC0517-5G
APOSPC0517-250MG
3-(Trifluoromethoxy)-DL-phenylglycine
3-(Trifluoromethoxy)-DL-phenylglycine
Formel:
C₉H₈F₃NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631460 CAS-Nummer: 261952-23-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...