3,4,5-Trifluoro-DL-phenylglycine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-2-(3,4,5-trifluorophenyl)acetic acid, H-DL-Phg(3,4,5-TriF)-OH

PC0533-1G PC0533-250MG PC0533-5G
APOSPC0533-1GEA 43.5 EUR
APOSPC0533-1G APOSPC0533-250MG APOSPC0533-5G
3,4,5-Trifluoro-DL-phenylglycine
3,4,5-Trifluoro-DL-phenylglycine
Formel: C₈H₆F₃NO₂
Molecular Weight: 205,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631415
CAS-Nummer: 261952-27-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR