Sodium trifluoromethanesulphinate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Trifluoromethanesulphinic acid Sodium Salt, Langlois Reagent
PC0579-5G
PC0579-25G
PC0579-100G
APOSPC0579-5GEA
21.9
EUR
APOSPC0579-5G
APOSPC0579-25G
APOSPC0579-100G
Sodium trifluoromethanesulphinate 95%
Sodium trifluoromethanesulphinate
Formel:
CHF₃O₂S Molecular Weight: 156,05 g/mol Schmelzpunkt: 350 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03092989 CAS-Nummer: 2926-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...