2'-Hydroxy-5'-(trifluormethoxy)acetophenon

Lieferant: Apollo Scientific
Achtung

PC0592-250MG PC0592-1G
APOSPC0592-250MGEA 21.9 EUR
APOSPC0592-250MG APOSPC0592-1G
2'-Hydroxy-5'-(trifluormethoxy)acetophenon
2'-Hydroxy-5'-(trifluormethoxy)acetophenon
Formel: C₉H₇F₃O₃
Molecular Weight: 220.15 g/mol
Schmelzpunkt: 28…31 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01091002
CAS-Nummer: 146575-64-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR