1-Chlor-2-fluor-3-nitrobenzol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chloro-2-fluoronitrobenzene
PC063-5G
PC063-25G
PC063-1G
APOSPC063-5GEA
16.1
EUR
APOSPC063-5G
APOSPC063-25G
APOSPC063-1G
1-Chlor-2-fluor-3-nitrobenzol 98%
1-Chloro-2-fluoro-3-nitrobenzene
Formel:
C₆H₃ClFNO₂ Molecular Weight: 175,55 g/mol Siedepunkt: 106 °C/5mmHg Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00069417 CAS-Nummer: 2106-49-2 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...