2-Ethoxy-5-fluoro-4-pyrimidinone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Fluorouracil Related Compound F, 2-Ethoxy-5-fluoropyrimidin-4(3H)-one, 5-Fluoro-2-ethoxy-4-pyrimidinone
PC0704-25G
APOSPC0704-25GEA
65.4
EUR
APOSPC0704-25G
2-Ethoxy-5-fluoro-4-pyrimidinone
2-Ethoxy-5-fluoro-4-pyrimidinone
Formel:
C₆H₇FN₂O₂ Molecular Weight: 158,13 g/mol Schmelzpunkt: 180…184 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03265436 CAS-Nummer: 56177-80-1 EINECS: 260-029-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...