4-(4-Fluorophenyl)butyric acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(4-Fluorophenyl)butyric Acid
PC0715-5G
PC0715-1G
APOSPC0715-5GEA
270
EUR
APOSPC0715-5G
APOSPC0715-1G
4-(4-Fluorophenyl)butyric acid 98%
4-(4-Fluorophenyl)butyric acid
Formel:
C₁₀H₁₁FO₂ Molecular Weight: 182,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788503 CAS-Nummer: 589-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...