4-Brom-2,6-difluorbenzoesäure

Lieferant: Apollo Scientific
Achtung

PC0737-100G PC0737-5G PC0737-25G
APOSPC0737-100GEA 210 EUR
APOSPC0737-100G APOSPC0737-5G APOSPC0737-25G
4-Brom-2,6-difluorbenzoesäure
4-Bromo-2,6-difluorobenzoic acid
Formel: C₇H₃BrF₂O₂
Molecular Weight: 237 g/mol
Schmelzpunkt: 198…204 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094085
CAS-Nummer: 183065-68-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR