4-Fluor-m-toluonitril 98%

Lieferant: Apollo Scientific
Achtung

PC0774-10G PC0774-50G
APOSPC0774-10GEA 58.2 EUR
APOSPC0774-10G APOSPC0774-50G
4-Fluor-m-toluonitril 98%
4-Fluor-m-toluonitril
Formel: C₈H₆FN
Molecular Weight: 135,14 g/mol
Schmelzpunkt: 53…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788510
CAS-Nummer: 185147-08-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR