6-Fluorothio-4-chromanone 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Fluorothiochroman-4-one

PC0776-25G PC0776-5G
APOSPC0776-25GEA 244 EUR
APOSPC0776-25G APOSPC0776-5G
6-Fluorothio-4-chromanone 97%
6-Fluorothio-4-chromanone
Formel: C₉H₇FOS
Molecular Weight: 182,22 g/mol
Schmelzpunkt: 89…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00078488
CAS-Nummer: 21243-18-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR