4-Iodphenylsulfurpentafluorid
Lieferant: Apollo Scientific
|
Achtung
|
PC0790-1G
PC0790-5G
PC0790-10G
APOSPC0790-1GEA
194
EUR
APOSPC0790-1G
APOSPC0790-5G
APOSPC0790-10G
4-Iodphenylsulfurpentafluorid
4-Iodphenylsulfurpentafluorid
Formel:
C₆H₄F₅IS Molecular Weight: 330,600 g/mol Siedepunkt: 50 °C/0,75mmHg Schmelzpunkt: 36 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03788514 CAS-Nummer: 286947-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...