3-(Pentafluorthio)benzonitril 97%

Lieferant: Apollo Scientific
Achtung

PC0803-1G PC0803-5G
APOSPC0803-1GEA 400 EUR
APOSPC0803-1G APOSPC0803-5G
3-(Pentafluorthio)benzonitril 97%
3-(Pentafluorthio)benzonitril
Formel: C₇H₄F₅NS
Lagertemperatur: Raumtemperatur
MDL: MFCD03788520
CAS-Nummer: 401892-82-8
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR