O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
PFBHA.HCl
PC0810-1G
PC0810-5G
PC0810-25G
APOSPC0810-1GEA
65.4
EUR
APOSPC0810-1G
APOSPC0810-5G
APOSPC0810-25G
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride 97%
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride
Derivatising agent for Electron-Capture detection of steriods.
Formel:
C₇H₄F₅NO Molecular Weight: 249.57 g/mol Schmelzpunkt: 227 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012953 CAS-Nummer: 57981-02-9 EINECS: 261-057-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...