4-(Pentafluorothio)benzoyl chloride 97+%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(Pentafluorosulphanyl)benzoyl chloride

PC0813-1G PC0813-5G PC0813-25G
APOSPC0813-1GEA 290 EUR
APOSPC0813-1G APOSPC0813-5G APOSPC0813-25G
4-(Pentafluorothio)benzoyl chloride 97+%
4-(Pentafluorothio)benzoyl chloride
Formel: C₇H₄ClF₅OS
Molecular Weight: 266.61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03788523
CAS-Nummer: 197384-98-8
UN: 1760
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR