4-Fluorsalicylaldehyd 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluoro-2-hydroxybenzaldehyde

PC0836-1G PC0836-25G PC0836-5G
APOSPC0836-1GEA 21.9 EUR
APOSPC0836-1G APOSPC0836-25G APOSPC0836-5G
4-Fluorsalicylaldehyd 99%
4-Fluorsalicylaldehyd
Formel: C₇H₅FO₂
Molecular Weight: 140,1118 g/mol
Schmelzpunkt: 70…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788526
CAS-Nummer: 348-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR