2-Methoxy-5-(trifluormethyl)benzaldehyd 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC0840-5G
PC0840-25G
PC0840-100G
APOSPC0840-5GEA
55.1
EUR
APOSPC0840-5G
APOSPC0840-25G
APOSPC0840-100G
2-Methoxy-5-(trifluormethyl)benzaldehyd 97%
2-Methoxy-5-(trifluoromethyl)benzaldehyde
Formel:
C₉H₇F₃O₂ Molecular Weight: 204.15 g/mol Schmelzpunkt: 85…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788528 CAS-Nummer: 146539-83-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...