3,5-Difluorophenyl isocyanate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,3-Difluoro-5-isocyanatobenzene
PC0845-1G
PC0845-25G
PC0845-5G
APOSPC0845-1GEA
21.9
EUR
APOSPC0845-1G
APOSPC0845-25G
APOSPC0845-5G
3,5-Difluorophenyl isocyanate
3,5-Difluorophenyl isocyanate
- May form small amount of sediment.
Formel:
C₇H₃F₂NO Molecular Weight: 155.104 g/mol Siedepunkt: 159 °C Dichte: 1.31 g/cm³ (25 °C) Flammpunkt: 54 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02093693 CAS-Nummer: 83594-83-6 UN: 3080 ADR: 6.1+3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...