(3-Chloro-4-fluorophenyl)thiourea

Lieferant: Apollo Scientific
Gefahr

PC0851-1G PC0851-5G
APOSPC0851-1GEA 68.3 EUR
APOSPC0851-1G APOSPC0851-5G
(3-Chloro-4-fluorophenyl)thiourea
(3-Chloro-4-fluorophenyl)thiourea
Formel: C₇H₆ClFN₂S
Molecular Weight: 204.65 g/mol
Schmelzpunkt: 236 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01571081
CAS-Nummer: 154371-25-2
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR