1-(Brommethyl)-3-(4-fluorphenoxy)benzol 97%
Lieferant: Apollo Scientific
Synonyme:
3'-Bromomethyl-4-fluorodiphenyl Ether
PC0853-1G
PC0853-5G
APOSPC0853-1GEA
328
EUR
APOSPC0853-1G
APOSPC0853-5G
1-(Brommethyl)-3-(4-fluorphenoxy)benzol 97%
3-(4-Fluorophenoxy)benzyl bromide
Formel:
C₁₃H₁₀BrFO Molecular Weight: 281.12 g/mol Siedepunkt: 160 °C/2mmHg Lagertemperatur: Kühlschrank |
MDL:
MFCD00059915 CAS-Nummer: 65295-58-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...