2-Chlor-4-fluor-5-nitroanisol

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Chloro-5-fluoro-2-methoxy-4-nitrobenzene

PC0858-1G PC0858-5G PC0858-25G
APOSPC0858-1GEA 21.9 EUR
APOSPC0858-1G APOSPC0858-5G APOSPC0858-25G
2-Chlor-4-fluor-5-nitroanisol
2-Chlor-4-fluor-5-nitroanisol
Formel: C₇H₅ClFNO₃
Molecular Weight: 205,5700 g/mol
Schmelzpunkt: 72…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03425810
CAS-Nummer: 84478-76-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR