2-Fluor-6-methoxybenzoylchlorid

Lieferant: Apollo Scientific
Gefahr

PC0868-1G PC0868-5G
APOSPC0868-1GEA 42.2 EUR
APOSPC0868-1G APOSPC0868-5G
2-Fluor-6-methoxybenzoylchlorid
2-Fluor-6-methoxybenzoylchlorid
Formel: C₈H₆ClFO₂
Molecular Weight: 188.59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03788536
CAS-Nummer: 500912-12-9
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR