2-Amino-5-fluorpyridin 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC0886-5G
PC0886-10G
PC0886-1G
APOSPC0886-5GEA
26.1
EUR
APOSPC0886-5G
APOSPC0886-10G
APOSPC0886-1G
2-Amino-5-fluorpyridin 99%
2-Amino-5-fluorpyridin
Formel:
C₅H₅FN₂ Molecular Weight: 112,11 g/mol Schmelzpunkt: 93…99 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861120 CAS-Nummer: 21717-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...