2'-Chlor-6'-fluoracetophenon 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2-Chloro-6-fluorophenyl)ethanone
PC0902-250MG
PC0902-25G
PC0902-5G
PC0902-100G
APOSPC0902-250MGEA
21.9
EUR
APOSPC0902-250MG
APOSPC0902-25G
APOSPC0902-5G
APOSPC0902-100G
2'-Chlor-6'-fluoracetophenon 97%
2'-Chlor-6'-fluoracetophenon
Formel:
C₈H₆ClFO Molecular Weight: 172.59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631390 CAS-Nummer: 87327-69-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...