2-Brom-4,5,6-trichloranilin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromo-2,3,4-trifluoroaniline, 2-Bromo-4,5,6-trifluoroaniline
PC0936-1G
PC0936-5G
PC0936-10G
APOSPC0936-1GEA
21.9
EUR
APOSPC0936-1G
APOSPC0936-5G
APOSPC0936-10G
2-Brom-4,5,6-trichloranilin 98%
2-Brom-4,5,6-trichloranilin
Formel:
C₆H₃BrF₃N Molecular Weight: 225.9900 g/mol Schmelzpunkt: 58…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013388 CAS-Nummer: 122375-82-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...