1-Brom-3-fluor-4-nitrobenzol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Brom-2-fluornitrobenzol

PC0939-100G PC0939-25G PC0939-5G
APOSPC0939-100GEA 114 EUR
APOSPC0939-100G APOSPC0939-25G APOSPC0939-5G
1-Brom-3-fluor-4-nitrobenzol 97%
1-Bromo-3-fluoro-4-nitrobenzene
Formel: C₆H₃BrFNO₂
Molecular Weight: 220 g/mol
Schmelzpunkt: 84…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01930221
CAS-Nummer: 321-23-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR